Learn More
Research Products International Corp Culture tubes, Polypropylene, Non-graduated, 17x100 mm, S w/2 Position Cap, 1000/cs
SDP
Supplier:  Research Products International Corp 168599LC
Encompass
These culture tubes are manufactured from premium quality polystyrene or polypropylene. Tubes are offered in two sizes: 12 x 75mm (nominal capacity 5ml) and 17 x 100mm (nominal capacity 15ml). Tubes are supplied sterile, capped with a two position polyethylene closure that can be adjusted for aerobic or anaerobic applications. Correct cap position is achieved with an easy one hand operation. Supplied in 8 bags of 125 tubes.
Catalog No. 50-222-9683