Learn More
Selleck Chemical LLC Culmerciclib E4703-5MG
SDP
Supplier:  Selleck Chemical LLC E47035MG
Encompass_Preferred
Culmerciclib (TQB3616) is a potent inhibitor of cyclin dependent kinase (CDK)4/6 and exhibits highly effective antineoplastic activity TQB3616 exhibits significant synergistic antitumor activity in estrogen receptor (ER)-positive/HER2-negative or HER2-positive breast cancer when combined with endocrine therapy or Human Epidermal Growth Factor Receptor 2 (HER2) targeted therapy
Catalog No. 50-000-43629