Learn More
Lonza Walkersville TheraPRO CHO Cloning Medium 1L
Supplier:  Lonza Walkersville BESP1609Q
Encompass
TheraPRO CHO Cloning Medium supports the growth and viability of GS-CHO cells during a variety of cloning methods in the cell line development process This chemically defined medium is ideal for isolating clones in addition to supporting clone recovery during the stressful and crucial process All components of the TheraPRO CHO Media System are manufactured in ISO 13485 certified facilities in accordance with GMP and are accompanied by a Drug Master File (DMF) and assistance with regulatory submissions
Catalog No. 50-004-96335