Learn More
Bellco Glass, Inc. Culture Tube Rack,16mm
SDP
Supplier:  Bellco Glass, Inc. 202816064
Encompass_Preferred
The culture tube rack is designed for roller culture tubes. Manufactured from 303 stainless steel construction. Includes spring clips that prevent tubes from moving in the rack. SST WIRE RACK FOR 38mm OD TUBES, SPACING: 2x6 - HOLDS 12 TUBES, DIMENSIONS: 10" WIDE X 3 1/2" DEEP X 2 7/8" TALL, EACH OPENING IN RACK: 1 9/16" (39.6mm). MADE IN THE USA. WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .162"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each
Catalog No. 50-194-5325