Learn More
FUJIFILM BIOSCIENCES INC CultureSure IWR-1-endo
Supplier:  FUJIFILM BIOSCIENCES INC 03325133
Encompass_Preferred
This is a Wnt signaling inhibitor IWR-1 stabilizes a complex consisting of Axin2 Apc Ck1 and Gsk3b that degrades beta catenin and promotes decomposition of beta catenin
Catalog No. 50-004-50246