Learn More
FUJIFILM BIOSCIENCES INC CultureSure Monosodium L-Gluta
Supplier:  FUJIFILM BIOSCIENCES INC 03524135
Encompass_Preferred
High-purity glutamate monohydrate is suitable for sensitive cell culture systems This product has been examined by endotoxins testing and mycoplasma testing Our culture medium has been used successfully and can be used safely
Catalog No. 50-004-50265