Learn More
FUJIFILM BIOSCIENCES INC CultureSure PD0325901
Supplier:  FUJIFILM BIOSCIENCES INC 03226183
Encompass_Preferred
PD0325901 is a MAPK inhibitor with potent antitumor activity It induces phosphorylation of MAPK inhibition of its activation and inhibition of tumor cell proliferation PD0325901 inhibits MAPK activity without adverse reactions Addition of this product and CHIR99021 to a culture medium results in efficient culture of mouse ES cells It has been reported that use of this product with SB431542 and thiazovivin not only improves reprogramming efficiency 200 folds or more but also accelerates reprogramming speed
Catalog No. 50-004-50314