Learn More
American Radiolabeled Chemicals Inc CUMI 101 3H 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 175550UC
Encompass_Preferred
CUMI-101-methyl-3H 50uCi in ethanol, 1 mCi/ml, S.A. 70-87 Ci/mmol, M.W. 373.45 Shipped in Dry Ice, Exclusive
Catalog No. 50-901-188