Learn More
ATCC Cupriavidus metallidurans; Strain: CH34
Supplier:  ATCC 43123
Encompass
Formerly Wautersia metallidurans. Genome sequencing strain. Type strain. Resistant to zinc, cobalt, nickel, mercury, cadmium, and produces a zinc-binding protein. Purified genomic DNA of this strain is available as ATCC 43123D-5™.
Catalog No. 50-238-2989