Learn More
Selleck Chemical LLC Curcumin S1848-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S184810MM/1ML
Encompass_Preferred
Curcumin (Diferuloylmethane Natural Yellow 3 Turmeric yellow) is the principal curcuminoid of the popular Indian spice turmeric which is a member of the ginger family (Zingiberaceae) It is an inhibitor of p300 histone acetylatransferase IC50 25 M and Histone deacetylase (HDAC) activates Nrf2 pathway and supresses the activation of NF- B Curcumin induces mitophagy autophagy apoptosis and cell cycle arrest with antitumor activity Curcumin reduces renal damage associated with rhabdomyolysis by decreasing ferroptosis-mediated cell death Curcumin exhibits anti-infective properties against various human pathogens like the influenza virus hepatitis C virus HIV and so on
Catalog No. 50-311-0571