Learn More
Medchemexpress LLC Curcumin analogue | 1258513-40-4 | 99.2% | C18H17N3O | 10MG
SDP
Supplier:  Medchemexpress LLC HY13570210MG
Encompass_Preferred
UBS109 is a curcumin analogue used in bone research that promotes osteoblast differentiation and mineralization by activating Smad signaling and inhibiting NF-κB. It is supplied as a high-purity research chemical suitable for in vitro studies in bone tissue engineering and osteoporosis models.
  • Promotes osteoblast differentiation and mineralization
  • Activates Smad signaling and inhibits NF-κB
  • Suitable for bone tissue engineering and osteoporosis research
  • High purity (99.2%) for research applications
  • Available in small research pack sizes (10 mg)
Catalog No. 50-002-60822