Learn More
Medchemexpress LLC Curcumin-β-D-glucuronide | 227466-72-0 | 98.01% | 544.50 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY138795
Encompass_Preferred
Curcumin-β-D-glucuronide is a major metabolite after oral intake of Curcumin in hepatic tissue and portal blood. It can be used for the research of colon cancer.
  • Exhibits potent anti-tumor effects on KRASm/TP53w HCT116 xenograft model
  • Suppresses tumor growth by 38%
  • Does not show a negative impact on body weight
  • Attenuates NF-κB activity
  • Appears as a yellow to brown solid
  • Soluble in DMSO: 50 mg/mL (91.83 mM; ultrasonic and warming and heat to 60°C)
Catalog No. 50-003-50487