Learn More
Medchemexpress LLC Curcumin-β-d-glucuronide | 227466-72-0 | MFCD28963761 | 98.0% | 544.50 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13879510MM/1ML
Encompass_Preferred
Curcumin-β-d-glucuronide is a major metabolite after oral intake of Curcumin, found in hepatic tissue and portal blood. It is used for research on colon cancer and has demonstrated anticancer properties in mouse tumor models and human metastatic colorectal cancer cell lines by promoting apoptosis. It also shows potential as a colon cancer prevention agent and possesses anti-inflammatory attributes by inhibiting prostaglandin E2 (PGE2) levels.
  • Major metabolite of curcumin
  • Exhibits anticancer properties
  • Promotes apoptosis in cancer cells
  • Potential colon cancer prevention agent
  • Possesses anti-inflammatory attributes
  • Inhibits prostaglandin E2 (PGE2) levels
Catalog No. 50-003-52385