Learn More
Fine Science Tools CURVED 1 X 2 TEETH 14 CM
SDP
Supplier:  Fine Science Tools 1102914
Encompass
Tip Shape: Curved/ Tips: 1x2 Teeth/ Tip Dimensions: 3 x 2mm/ Alloy / Material: Stainless Steel/ Length: 14cm
Catalog No. NC9123118