Learn More
Fine Science Tools CURVED MAYO SCISSORS 15CM
SDP
Supplier:  Fine Science Tools 1411115
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 39mm/ Alloy / Material: Stainless Steel/ Length: 15cm/ Serrated: Yes/ Feature: ToughCut
Catalog No. NC1670589