Learn More
Cayman Chemical CurvularIa CurvulIn 5mg
SDP
Supplier:  Cayman Chemical 196095 MG
Encompass_Preferred
A phytotoxin reported to inhibit microtubule assembly and iNOS expression
Catalog No. 50-298-0639