Learn More
Medchemexpress LLC Curzerenone | 20493-56-5 | 5 MG

Supplier: Medchemexpress LLC HYN36515MG
Curzerenone is a constituent of leaf essential oil extracted from *L. pulcherrima* and shows slight inhibitory effectiveness against *E. coli*. This compound is supplied as a colorless to light yellow liquid with high purity.
- Purity of 98.55%
- Molecular formula: C15H18O2
- Molecular weight: 230.30
- Soluble in DMSO at 50 mg/mL
- Store at -20°C, protected from light
- In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.