Learn More
Aldevron Custom Megaprep, KanR, Tris
Supplier:  Aldevron FISHERKANMEGA_T
Encompass
Take the burden of prepping plasmids off your hands by sending your plasmid preps to Aldevron. In as fast as 72 hours from receipt of your starting material, we will make a 0.5L batch scale of your plasmid and deliver all plasmid purified from that scale, typically yielding approximately 4mg* delivered. You will receive your Kanamycin-resistant plasmid delivered in Tris buffer (10mM Tris, pH 8.0) with UV Spec QC.
To initiate this project, you can add the quantity of plasmids required to your Fisher Cart.
REQUIRED:
Download this submission form and send the completed form to order@aldevron.com. We’ll be able to get your project started upon receipt of your starting materials as described in the form. Starting Material Submission Form
*Note: plasmid yield not guaranteed, yield estimate provided for information only
Catalog No. NC3869413