Learn More
World Precision Instrument Custom MicroFil-CMF90U
SDP
Supplier:  World Precision Instrument CMF90U
Encompass
WPI's MicroFil flexible plastic needles fills micropipettes easily and reliably. Its long and fine tip allows you to start the filling very close to the pipette tip, eliminating both air bubble formation and clogging due to the washing down of dust particles. The transparent amber MicroFil needle is constructed from a combination of plastic and fused silica - no metal components are used. The MicroFil needle can be stored for days with the filling solution inside without clogging. Sold in packages of 4.
  • This is a customized item; please specify the length needed in whole centimeters, up to 50cm
  • Non-returnable, non-refundable item
Catalog No. 50-253-8887