Learn More
Grainger Cut Off Blade 3/16 Carbide Tip
Supplier:  Grainger 16T526
Encompass_Preferred
The Micro 100 Indexable Chamfering Tool is designed for high-performance chamfering and facing. It has a 16mm shank with Weldon flat for secure holding and uses triangular TiAlN-coated inserts for wear resistance on various materials. It produces chamfers from 0.04" to 0.24" efficiently in one pass. Durable carbide inserts ensure long life and reduced tool change costs. Its design allows easy insert changes, minimizing downtime, making it a cost-effective chamfering solution.
Catalog No. 50-276-6005