Learn More
Grainger Cut Off Blade 3/16 Carbide Tip
Supplier:  Grainger 16T530
Encompass_Preferred
The Grainger 16T530 is a Dayton 1/3 HP 115/230V Dual Voltage Farm Duty Electric Motor. It is a general purpose motor with a 1/3 horsepower rating and operates on either 115 or 230 volt power. The motor has a 1075 RPM operating speed with a 56C frame size. It features permanent split capacitor construction and is thermally protected. Applications include driving pumps, fans, and shop tools. The motor dimensions are 7-1/2" diameter x 8-3/4" long. It comes with a 12 month warranty from Grainger.
Catalog No. 50-276-5572