Learn More
Grainger Cut Off Blade 5/32 Carbide Tip
Supplier:  Grainger 16T519
Encompass_Preferred
The Grainger 16T519 is a 16-ton hydraulic bottle jack made of steel with a lifting range of 10-15/16 inches to 16-1/2 inches. It has a built-in overload valve and bypass device for controlled lifting. The jack's lift saddle tilts and rotates to accommodate any shape load. It has a weight capacity of 32,000 lbs and measures 9-3/8 x 5-1/8 x 8-3/4 inches. This hydraulic jack is designed for shop service, general maintenance work and farm equipment repair.
Catalog No. 50-276-5809