Learn More
Grainger Cut-Off GDM 2520N020PM PR1225 10 pcs.
Supplier:  Grainger 170FC9
Encompass_Preferred
The Grainger 170FC9 is a Dayton 1/3 HP Centrifugal Pump with a 115V motor that pumps up to 360 GPH. It has a thermoplastic body and impeller and a stainless steel shaft. The pump has a 1/2" NPT inlet and outlet and operates at 3450 RPM. It can handle temperatures up to 130°F and has a 6 foot power cord. The overall dimensions are 7-1/2" length x 5-1/2" width x 6-3/4" height. This centrifugal utility pump is designed for transferring water, light oils and chemicals.
Catalog No. 50-276-6146