Learn More
Grainger Cut-Off GDM 3020N025PM PR1225 10 pcs.
Supplier:  Grainger 170FC5
Encompass_Preferred
The Grainger 170FC5 is a 5-gallon plastic utility pail made from high-density polyethylene. It features a wide opening for easy filling, a bail handle for carrying, and gallon increments for measuring. Suitable for mixing, transporting, and waste collection, it meets FDA food contact regulations. With an impact rating of -20°F to 120°F, it has a 5-year shelf life, weighs 2.2 lbs, and measures 12" diameter x 11.5" height. Ideal for commercial, industrial, and food service use.
Catalog No. 50-276-5496