Learn More
Grainger Cut-Off GDM 3020N025PM PR1535 10 pcs.
Supplier:  Grainger 170FV3
Encompass_Preferred
The 170FV3 is a 3-speed, 1/2 HP, 115V, single phase, totally enclosed fan cooled motor made by Century. It has a 48 frame size, measures 10-1/2" long, has a 1-1/8" diameter shaft, operates at 1725 RPM, and is thermally protected. This motor can be used for various industrial applications requiring a reliable, enclosed motor with moderate horsepower and speed. With proper maintenance, this Century electric motor should provide years of continuous service.
Catalog No. 50-276-4737