Learn More
Grainger Cut-Off GDM 3020R025PM6D PR1535 10 pcs.
Supplier:  Grainger 170FV6
Encompass_Preferred
The 170FV6 is a 1/2 HP, 115V, single phase, open drip proof fan motor manufactured by Century. It has a 1075 RPM operating speed and is thermally protected. The motor is constructed with an aluminum housing, has sleeve bearings, and is totally enclosed and fan cooled. It can be used with blowers, fans, and pumps in industrial and commercial HVAC applications. The motor dimensions are 9-1/4" long x 5-3/4" wide x 6-7/8" high and it weighs 15 pounds.
Catalog No. 50-276-3889