Learn More
Grainger Cut-Off TKF 12R100S16DR KW10a 10 pcs.
Supplier:  Grainger 170EW0
Encompass_Preferred
The Grainger 170EW0 is a Westinghouse 15 Amp, 125 Volt, 2 Pole, 3 Wire, Grounding NEMA 5-15R Commercial Grade Duplex Receptacle. It has side and back wire terminals and is UL listed with a high impact thermoplastic face. The receptacle is ivory in color and measures 4.19" x 2.25" x 1.62". It can be used for general purpose power connections.
Catalog No. 50-276-5791