Learn More
Grainger Cut-Off TKF 16R200S16DR PR1225 G 10 pcs.
Supplier:  Grainger 170FG9
Encompass_Preferred
The Grainger 170FG9 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features sleeve bearings, manual reset thermal overload protection, and meets NEMA design B specifications. This 1/3 HP motor can provide 0.25 kW of output power.
Catalog No. 50-276-6297