Learn More
Electron Microscopy Sciences Safeknit™ Cut-Resistant Gloves Heavy Duty, Size 8
SDP
Supplier:  Electron Microscopy Sciences 7106508
Encompass_Preferred
High performance cut resistant Safeknit glove is a blend of abrasion-resistant Spectra fiber and other synthetics. The mix of these fibers form a rugged cut resistant barrier that offers the histologist sensitivity for manipulation but with extreme comfort and productivity.
NOTE: These gloves are neither puncture proof nor water resistant. They should not be used to protect against intentional cutting. Avoid use near moving blades. Two strand seamless glove.
Size: 8
Catalog No. 50-930-743