Learn More
Active Motif CUT&Tag-IT R-loop Kit 16 rxns
SDP
Supplier:  Active Motif 53167
Encompass
It is easy to profile DNA-RNA hybrid R-loops genome-wide with Active Motif’s CUT&Tag-IT™ R-loop Assay Kit. The kit utilizes a CUT&Tag-based approach to profile R-loops genome-wide using a specifically optimized protocol for cell samples and our DNA-RNA hybrid (Clone S9.6) antibody. This is not simply the same as our CUT&Tag-IT Assay Kit with a DNA-RNA Hybrid mAb (Clone S9.6). The CUT&Tag-IT R-loop Assay Kit includes optimized DNA Binding Buffer, modified DNA Purification Columns, Q5 Polymerase Master Mix and a second mixing step during secondary antibody incubation all performed in an efficient strip-tube format protocol specifically designed for R-loop targets.
Catalog No. NC2586635