Learn More
Medchemexpress LLC Cutamesine dihydrochloride | 165377-44-6 | MFCD00939308 | 98.1% | 441.43 g/mol | C23H34Cl2N2O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY135105MG
Encompass_Preferred
Cutamesine dihydrochloride is a selective sigma-1 (σ1) receptor agonist supplied as the dihydrochloride salt for research use. It is provided as a solid with reported purity of ~98% and a molecular weight of 441.43 g/mol. The compound is used in neuroscience and pharmacology studies to probe sigma-1 receptor activity and downstream signaling pathways.
  • Selective sigma-1 receptor agonist for receptor pharmacology studies.
  • High purity suitable for research applications.
  • Supplied as a solid with reported solubility in DMSO and water.
  • Available in small pack sizes appropriate for screening and assay work.
  • Applicable to in vitro and ex vivo neuroscience models.
Catalog No. 50-002-21910