Learn More
World Precision Instrument Cutting Mats for Biopsy Punches-504621
SDP
Supplier:  World Precision Instrument 504621
Encompass
Use our cutting mats for clean cuts through your tissue samples. Our mats ensure that your cored sample comes out whole without burrs or rough edges. Our 1.5mm thick, five layer composite cutting mats are reversible, inert and self-healing. These mats are designed for use with your 
Catalog No. 50-253-9324