Learn More
Electron Microscopy Sciences EMS High Precision Cutting Tweezer Style 15AFW, 115 mm
SDP
Supplier:  Electron Microscopy Sciences 7832515AF
Encompass_Preferred
For cutting hair sprigs, electronic component wires, copper, gold, silver and other soft wires. Large head, oblique strong cut at the cutter's tip. Cutting edge is limited to the tip area. 111 mm long. M = 11 mm, N = 3 mm, P = 24 mm.
OAL: 115 mm
Catalog No. 50-193-0588