Learn More
Hach Company CuVer 2 Copper Reagent AccuVac Ampules, pk/25
Supplier:  Hach Company 2504025
Encompass
For determination of Copper by the Bicinchoninate method. Hach Method 8026. For copper in the presence of high levels of aluminum, calcium and iron. Range: 0.04 - 5.00 mg/L Cu. Pack of 25 ampules
Catalog No. 50-199-6102
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.