Learn More
Fireflysci A23 Cell Mount (For Type 4)
SDP
Supplier:  Fireflysci A23
Encompass_Preferred
Adapter for allowing Type 4 cuvettes to fit into a standard 10mm cuvette holder• Dimensions- 12.5x12.5x40mm
Catalog No. 50-106-0494