Learn More
PERFECTOR SCIENTIFIC Disposable Cuvette, Standard, 3.5mL, 340-800nm, 500/pack
Supplier:  PERFECTOR SCIENTIFIC 9002
Encompass
All cuvettes have a 10mm path length and are 45mm high. Internal width of the semi-micro size is 4mm. Fluorometer cuvettes have all four sides clear. Cuvettes for use in the visible spectra are made from polystyrene, UV cuvettes are made from methacrylate. The cuvettes will fit all standard instruments The cuvettes are packed in 5 styrofoam trays, each tray holding 100 cuvettes. The trays have a tight fitting styrofoam lid which protects the optical surfaces and keeps cuvettes clean.
Catalog No. NC1016683