Learn More
Cayman Chemical CuZn SOD MoncLNal ANTIBDY
SDP
Supplier:  Cayman Chemical 37087100 UL
Encompass_Preferred
A neuropeptide fragment of ProSAAS and a GPR171 agonist selectively increases intracellular calcium levels in CHO cells expressing GPR171 over those expressing GPR19 GPR108 or GPR165 at 1 UM increases neurite outgrowth in mouse Neuro2a cells at 10UM
Catalog No. 50-291-5225