Learn More
Sino Biological Recombinant Coxsackievirus(CV)A6 VP1 Protein (Fc Tag)
Supplier:  Sino Biological 41053V02B100
Encompass_Preferred
A DNA sequence encoding the Coxsackievirus(CV)A6 VP1 (AHF49552.1) (Gly1-Met298) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-299-3347