Learn More
Electron Microscopy Sciences Tanner Coverglass Rectangle #1, 24 x 30 mm 10 OZ
SDP
Supplier:  Electron Microscopy Sciences 7188459
Encompass_Preferred
Tanner coverglass is suitable for a host of laboratory applications ranging from histopathology, cytology, microbiology and life sciences. It is ultrathin and disposable
Thickness: #1
Size: 24 x 30 mm
Catalog No. 50-192-8941