Learn More
Apexbio Technology LLC CVT 10216 1005334-57-5 25mg
SDP
Supplier:  Apexbio Technology LLC B492825
Encompass_Preferred
CVT 10216 (CAS 1005334-57-5) is a selective and reversible inhibitor of aldehyde dehydrogenase 2 (ALDH-2) exhibiting an IC50 value of 29 nM It also inhibits ALDH-1 though with lower potency (IC50 1 3 M) By modulating ALDH enzymatic activity CVT 10216 has been shown in animal studies to reduce ethanol consumption and display anxiolytic-like effects This compound is primarily utilized to investigate the involvement of ALDH-mediated pathways in the regulation of alcohol-related behaviors and anxiety supporting its use in studies of neurobehavioral and addiction mechanisms
Catalog No. 50-000-90408