Learn More
Selleck Chemical LLC CVT-313
SDP
Supplier:  Selleck Chemical LLC S653725MG
Encompass_Preferred
CVT-313 is a potent CDK2 inhibitor with an IC50 of 0 5 microM in vitro It has no effect on other nonrelated ATP-dependent serine/threonine kinases
Catalog No. 50-000-02661