Learn More
Selleck Chemical LLC CW069 2mg 1594094-64-0
SDP
Supplier:  Selleck Chemical LLC S73362MG
Encompass_Preferred
CW069 is an allosteric, and selective inhibitor of microtubule motor protein HSET with IC50 of 75 ?M, significant selectivity over KSP. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4662