Learn More
Selleck Chemical LLC CX-5461 5mg 1138549-36-6
SDP
Supplier:  Selleck Chemical LLC S26845MG
Encompass_Preferred
CX-5461 is an inhibitor of rRNA synthesis, selectively inhibits Pol I-driven transcription of rRNA with IC50 of 142 nM in HCT-116, A375, and MIA PaCa-2 cells, has no effect on Pol II, and possesses 250- to 300-fold selectivity for inhibition of rRNA transcription versus DNA replication and protein translation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2758