Learn More
Selleck Chemical LLC CX-6258 HCl
SDP
Supplier:  Selleck Chemical LLC S7041100MG
Encompass_Preferred
CX-6258 HCl is a potent orally efficacious pan-Pim kinase inhibitor with IC50 of 5 nM 25 nM and 16 nM for Pim1 Pim2 and Pim3 respectively
Catalog No. 50-000-02511