Learn More
Molecular Dimensions CX100 CRYOPAK DRY SHIPPER
SDP
Supplier:  Molecular Dimensions MD721
Encompass
Protein Crystallography. Dry Shipper (CX100) from Taylor-Wharton, designed to transport a variety of materials safely at cryogenic temperatures. Meets International Air Transport Association requirements. 4.4 L capacity. Lockable hinged lid. MEETS IATA (International Air Transport Association) requirements. Advanced concept absorbent enables faster charging. 4.4 L capacity. Compact, lightweight design. Optional hard shell shipping case (MD7-22) has shock resistant foam liner provides extra protection in transit. Lockable hinged lid.
Catalog No. NC9879599