Learn More
TARGETMOL CHEMICALS INC CX614 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1969825MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.CX614 is a positive allosteric modulator of the AMPA receptor concerning AMPA. Purity 97.43%
Catalog No. 50-287-8277