Learn More
Selleck Chemical LLC CY-09
SDP
Supplier:  Selleck Chemical LLC S577425MG
Encompass_Preferred
CY-09 is a specific NLRP3 inflammasome inhibitor that directly targeted NLRP3 itself It is tested against the five major cytochrome P450 enzymes 1A2 2C9 2C19 2D6 and 3A4 with half maximal inhibitory concentration (IC50) values of 18 9 8 18 50 50 and 26 0 M respectively
Catalog No. 50-000-03420