Learn More
TARGETMOL CHEMICALS INC CY3 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T189455MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. CY3 (Sulfo-Cyanine3) is an orange-fluorescent label for nucleic acids and proteins (λex=554, λem=568).
Catalog No. 50-287-8254