Learn More
Apexbio Technology LLC Cy3 carboxylic acid (non-sulfonated) 25mg
SDP
Supplier:  Apexbio Technology LLC A813225
Encompass_Preferred
Cy3 carboxylic acid (non-sulfonated) is a non-reactive fluorescent dye with excitation/emission wavelengths around 550/570 nm commonly used in biological imaging and assay calibration experiments Lacking a reactive group it does not directly bind biomolecules but can be conjugated through activation methods Due to limited aqueous solubility dissolution in organic solvents is typically required prior to biomolecular labeling In prior studies this dye has been conjugated to single-stranded DNA probes immobilized onto solid supports serving as donor fluorophores in FRET-based assays and supramolecular interaction studies such as monitoring metabolic substrate uptake at the single-cell level
Catalog No. 50-000-61685