Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cy3-Cisplatin | 85% | 50 MG
SDP

Supplier:  Medchemexpress LLC HYD245250MG

Encompass_Preferred

Cy3-Cisplatin is a compound that combines Cyanine 3 fluorescent dye (CY3) with Cisplatin. It is intended for research use only and can be utilized for drug delivery, fluorescent labeling, cell imaging, and tracking. The compound appears as an orange to red solid.

  • Combines Cyanine 3 fluorescent dye and Cisplatin
  • Used for drug delivery
  • Used for fluorescent labeling
  • Used for cell imaging and tracking
  • Appears as an orange to red solid
  • Store at -20°C, protect from light
  • Unstable in solutions, freshly prepared solutions are recommended

Catalog No. 50-004-31149


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.